Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3hcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146012.2 | NP_001139484.2 | 1 - 2 | 83 | 53 | |
GenBank mRNA | AK032854.1 | - | 1 - 2 | 53 | 53 | |
AK156843.1 | - | 1 - 2 | 53 | 53 | ||
CJ181933.1 | - | 1 - 2 | 80 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus 1700 Probe spans exons |