Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnaseh2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026001.2 | NP_080277.1 | 3 - 4 | 257 | 138 | |
GenBank mRNA | AK011914.1 | - | 3 - 4 | 296 | 138 | |
AK082752.1 | - | 3 - 4 | 258 | 138 | ||
BC047997.1 | - | 3 - 4 | 258 | 6510494 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |