Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp3ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293622.1 | NP_001280551.1 | 2 - 3 | 1033 | 111 | |
NM_008913.5 | NP_032939.1 | 2 - 3 | 1033 | 111 | ||
GenBank mRNA | AK146387.1 | - | 2 - 3 | 977 | 111 | |
AK150393.1 | - | 2 - 3 | 392 | 111 | ||
AK166287.1 | - | 2 - 3 | 810 | 111 | ||
AK209023.1 | - | 1 - 2 | 174 | 111 | ||
BC096652.1 | - | 2 - 3 | 275 | 111 | ||
BC138612.1 | - | 2 - 3 | 306 | 8861632 | 111 | |
J05479.1 | - | 2 - 3 | 330 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |