Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033350.2 | NP_001028522.2 | 2 - 3 | 580 | 100 | |
GenBank mRNA | AK041242.1 | - | 2 - 3 | 541 | 100 | |
AK156201.1 | - | 2 - 3 | 590 | 100 | ||
AY178734.1 | - | 2 - 3 | 600 | 100 | ||
BC026932.1 | - | 2 - 3 | 527 | 4008261 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |