Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038643.1 | NP_001033732.1 | 8 - 9 | 1882 | 64 | |
NM_023908.2 | NP_076397.2 | 8 - 9 | 1882 | 64 | ||
GenBank mRNA | AF226324.1 | - | 8 - 9 | 1780 | 64 | |
AK031280.1 | - | 8 - 9 | 1885 | 64 | ||
AK037938.1 | - | 8 - 9 | 1697 | 64 | ||
AK046945.1 | - | 7 - 8 | 1242 | 64 | ||
AK152488.1 | - | 8 - 9 | 1716 | 64 | ||
AK166266.1 | - | 8 - 9 | 1653 | 64 | ||
BC025059.1 | - | 8 - 9 | 1715 | 2598913 | 64 | |
BC094464.1 | - | 8 - 9 | 1674 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |