Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isg20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020583.5 | NP_065608.2 | 1 - 2 | 279 | 90 | |
GenBank mRNA | AK005528.1 | - | 1 - 2 | 71 | 90 | |
AK136756.1 | - | 1 - 2 | 205 | 90 | ||
BC022751.1 | - | 1 - 2 | 226 | 4912320 | 90 | |
BY087214.1 | - | 1 - 2 | 283 | 90 | ||
W97502.1 | - | 1 - 2 | 243 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |