Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntrk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008746.5 | NP_032772.3 | 10 - 11 | 1346 | 65 | |
NM_182809.2 | NP_877961.1 | 10 - 11 | 1346 | 65 | ||
GenBank mRNA | AF035399.1 | - | 11 - 12 | 1441 | 65 | |
AF035400.1 | - | 10 - 11 | 1495 | 65 | ||
AY336094.1 | - | 10 - 11 | 1291 | 65 | ||
BC139764.1 | - | 11 - 12 | 1358 | 40110345 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |