Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015795.1 | NP_056610.1 | 3 - 4 | 254 | 123 | |
GenBank mRNA | AF176531.1 | - | 3 - 4 | 254 | 123 | |
AK043554.1 | - | 3 - 4 | 224 | 123 | ||
AK081947.1 | - | 3 - 4 | 300 | 123 | ||
AK164657.1 | - | 3 - 4 | 337 | 123 | ||
BC086690.1 | - | 3 - 4 | 218 | 30683387 | 123 | |
BC115890.1 | - | 3 - 4 | 212 | 40103787 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |