Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081414.2 | NP_001074883.1 | 4 - 5 | 1632 | 106 | |
NM_001143834.1 | NP_001137306.1 | 4 - 5 | 1632 | 106 | ||
GenBank mRNA | AK032422.1 | - | 2 - 3 | 460 | 106 | |
BC096533.1 | - | 2 - 3 | 1017 | 106 | ||
BC144727.1 | - | 1 - 2 | 257 | 9053275 | 106 | |
EF508472.1 | - | 2 - 3 | 914 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |