Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081414.2 | NP_001074883.1 | 5 - 6 | 1858 | 124 | |
NM_001143834.1 | NP_001137306.1 | 5 - 6 | 1858 | 124 | ||
GenBank mRNA | BC096533.1 | - | 3 - 4 | 1243 | 124 | |
BC144727.1 | - | 2 - 3 | 483 | 9053275 | 124 | |
EF508472.1 | - | 3 - 4 | 1140 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |