Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081414.2 | NP_001074883.1 | 9 - 10 | 3341 | 76 | |
GenBank mRNA | AK032422.1 | - | 7 - 8 | 2169 | 76 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mus musculus 1700 Probe spans exons |