Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400016428.mm.1 |
NM_001253752.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016428.mm.1 |
NM_001253752.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016428.mm.1 |
NM_001253753.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016428.mm.1 |
NM_001253753.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016428.mm.1 |
NM_144842.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016428.mm.1 |
TC1400002132.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016429.mm.1 |
NM_001253752.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016429.mm.1 |
NM_001253752.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016429.mm.1 |
NM_001253753.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016429.mm.1 |
NM_001253753.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016429.mm.1 |
NM_144842.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016429.mm.1 |
TC1400002132.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016440.mm.1 |
NM_001253753.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016440.mm.1 |
NM_001253753.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016440.mm.1 |
NM_144842.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016440.mm.1 |
TC1400002132.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016441.mm.1 |
NM_001253753.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016441.mm.1 |
NM_001253753.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016441.mm.1 |
NM_144842.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016441.mm.1 |
TC1400002132.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016432.mm.1 |
NM_001253752.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016432.mm.1 |
NM_001253752.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016432.mm.1 |
NM_001253753.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016432.mm.1 |
NM_001253753.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016432.mm.1 |
NM_144842.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016432.mm.1 |
TC1400002132.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016434.mm.1 |
NM_001253752.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016434.mm.1 |
NM_001253752.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016434.mm.1 |
NM_001253753.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016434.mm.1 |
NM_001253753.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016434.mm.1 |
NM_144842.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016434.mm.1 |
TC1400002132.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016436.mm.1 |
NM_001253752.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016436.mm.1 |
NM_001253752.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016436.mm.1 |
NM_001253753.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016436.mm.1 |
NM_001253753.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016436.mm.1 |
NM_144842.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016436.mm.1 |
TC1400002132.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016442.mm.1 |
NM_001253753.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016442.mm.1 |
NM_001253753.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016442.mm.1 |
TC1400002132.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016435.mm.1 |
NM_001253752.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016435.mm.1 |
NM_001253752.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016435.mm.1 |
NM_001253753.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016435.mm.1 |
NM_001253753.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016435.mm.1 |
NM_144842.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016435.mm.1 |
TC1400002132.mm.1 |
MTA TC |