Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153458.3 | NP_703188.1 | 1 - 2 | 478 | 76 | |
GenBank mRNA | AF442825.1 | - | 1 - 2 | 193 | 76 | |
AK038569.1 | - | 1 - 2 | 425 | 76 | ||
AK044325.1 | - | 1 - 2 | 280 | 76 | ||
AK044605.1 | - | 1 - 2 | 428 | 76 | ||
AK045482.1 | - | 1 - 2 | 427 | 76 | ||
AK049612.1 | - | 1 - 2 | 479 | 76 | ||
AK079315.1 | - | 1 - 2 | 288 | 76 | ||
AK137414.1 | - | 1 - 2 | 280 | 76 | ||
BC112422.1 | - | 1 - 2 | 134 | 40061966 | 76 | |
BC113129.1 | - | 1 - 2 | 134 | 40061967 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |