Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010513.2 | NP_034643.2 | 5 - 6 | 1852 | 99 | |
GenBank mRNA | AF056187.1 | - | 5 - 6 | 1250 | 99 | |
AK155079.1 | - | 4 - 5 | 730 | 99 | ||
AK155884.1 | - | 4 - 5 | 748 | 99 | ||
BC138868.1 | - | 5 - 6 | 1282 | 8861890 | 99 | |
BC138869.1 | - | 5 - 6 | 1282 | 8861891 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |