Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243046.1 | NP_001229975.1 | 3 - 4 | 375 | 86 | |
NM_011807.3 | NP_035937.2 | 12 - 13 | 1630 | 86 | ||
GenBank mRNA | AK030221.1 | - | 3 - 4 | 376 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |