Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113471.1 | NP_001106942.1 | 5 - 6 | 640 | 90 | |
NM_175640.2 | NP_783571.2 | 5 - 6 | 696 | 90 | ||
GenBank mRNA | AK027962.1 | - | 2 - 3 | 356 | 90 | |
AK031445.1 | - | 4 - 5 | 621 | 90 | ||
AY161165.1 | - | 4 - 5 | 591 | 90 | ||
BC096685.1 | - | 5 - 6 | 641 | 1448913 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |