Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177740.5 | NP_808408.2 | 2 - 3 | 489 | 64 | |
GenBank mRNA | AJ557513.1 | - | 1 - 2 | 243 | 64 | |
AK145224.1 | - | 1 - 2 | 232 | 64 | ||
BC023870.1 | - | 1 - 2 | 297 | 5353958 | 64 | |
BC059072.1 | - | 2 - 3 | 489 | 6826181 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |