Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026043.3 | NP_080319.2 | 14 - 15 | 1656 | 134 | |
GenBank mRNA | BC016603.1 | - | 11 - 12 | 1172 | 3587176 | 134 |
BC043695.1 | - | 14 - 15 | 1625 | 5250774 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |