Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700012798.mm.1 |
NM_001159516.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012798.mm.1 |
NM_001159516.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012798.mm.1 |
NM_001159517.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012798.mm.1 |
NM_001159517.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012798.mm.1 |
NM_021881.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012798.mm.1 |
NM_021881.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012798.mm.1 |
TC1700001429.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012801.mm.1 |
NM_001159516.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012801.mm.1 |
NM_001159516.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012801.mm.1 |
NM_001159517.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012801.mm.1 |
NM_001159517.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012801.mm.1 |
NM_021881.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012801.mm.1 |
NM_021881.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012801.mm.1 |
TC1700001429.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012799.mm.1 |
NM_001159516.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012799.mm.1 |
NM_001159516.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012799.mm.1 |
NM_001159517.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012799.mm.1 |
NM_001159517.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012799.mm.1 |
NM_021881.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012799.mm.1 |
NM_021881.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012799.mm.1 |
TC1700001429.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012803.mm.1 |
NM_001159516.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012803.mm.1 |
NM_001159516.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012803.mm.1 |
NM_001159517.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012803.mm.1 |
NM_001159517.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012803.mm.1 |
NM_021881.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012803.mm.1 |
NM_021881.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012803.mm.1 |
TC1700001429.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012800.mm.1 |
NM_001159516.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012800.mm.1 |
NM_001159516.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012800.mm.1 |
NM_001159517.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012800.mm.1 |
NM_001159517.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012800.mm.1 |
NM_021881.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012800.mm.1 |
NM_021881.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012800.mm.1 |
TC1700001429.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012797.mm.1 |
NM_001159516.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012797.mm.1 |
NM_001159516.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012797.mm.1 |
NM_001159517.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012797.mm.1 |
NM_001159517.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012797.mm.1 |
NM_021881.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012797.mm.1 |
NM_021881.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012797.mm.1 |
TC1700001429.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012802.mm.1 |
NM_001159516.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012802.mm.1 |
NM_001159516.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012802.mm.1 |
NM_001159517.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012802.mm.1 |
NM_001159517.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012802.mm.1 |
NM_021881.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012802.mm.1 |
NM_021881.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012802.mm.1 |
TC1700001429.mm.1 |
MTA TC |