Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015795.1 | NP_056610.1 | 5 - 6 | 626 | 70 | |
GenBank mRNA | AF176531.1 | - | 5 - 6 | 626 | 70 | |
AK043554.1 | - | 5 - 6 | 596 | 70 | ||
AK164657.1 | - | 5 - 6 | 709 | 70 | ||
BC086690.1 | - | 5 - 6 | 590 | 30683387 | 70 | |
BC115890.1 | - | 5 - 6 | 584 | 40103787 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |