Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080443.2 | NP_536691.2 | 3 - 4 | 665 | 75 | |
NR_003961.1 | 4 - 5 | 740 | 75 | |||
GenBank mRNA | AK051017.1 | - | 4 - 5 | 653 | 75 | |
BC066824.1 | - | 2 - 3 | 167 | 30087227 | 75 | |
BC085155.1 | - | 3 - 4 | 645 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |