Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271402.1 | NP_001258331.1 | 1 - 2 | 265 | 72 | |
GenBank mRNA | AK002415.1 | - | 1 - 2 | 235 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |