Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gja3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271623.1 | NP_001258552.1 | 1 - 2 | 200 | 120 | |
GenBank mRNA | AY390399.1 | - | 1 - 2 | 200 | 120 | |
BQ554641.1 | - | 1 - 2 | 187 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |