Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008683.3 | NP_032709.1 | 2 - 3 | 161 | 74 | |
GenBank mRNA | AK146732.1 | - | 2 - 3 | 162 | 74 | |
AK147080.1 | - | 2 - 3 | 146 | 74 | ||
BC004625.1 | - | 2 - 3 | 146 | 3585834 | 74 | |
D10918.1 | - | 2 - 3 | 128 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |