Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034910.3 | NP_001030082.1 | 1 - 2 | 62 | 73 | |
GenBank mRNA | AK079042.1 | - | 1 - 2 | 63 | 73 | |
DQ012046.1 | - | 1 - 2 | 62 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |