Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem55b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033271.4 | NP_001028443.1 | 5 - 6 | 697 | 91 | |
GenBank mRNA | AK144251.1 | - | 5 - 6 | 697 | 91 | |
AK152640.1 | - | 5 - 6 | 683 | 91 | ||
AK152658.1 | - | 5 - 6 | 683 | 91 | ||
AK159641.1 | - | 5 - 6 | 701 | 91 | ||
AK169026.1 | - | 2 - 3 | 1541 | 91 | ||
AK205696.1 | - | 2 - 3 | 96 | 91 | ||
AK213231.1 | - | 2 - 3 | 96 | 91 | ||
AK219909.1 | - | 2 - 3 | 96 | 91 | ||
BC027355.1 | - | 5 - 6 | 682 | 91 | ||
BC039161.1 | - | 4 - 5 | 534 | 91 | ||
BC049945.1 | - | 4 - 5 | 554 | 91 | ||
BC058171.1 | - | 5 - 6 | 646 | 91 | ||
BC106163.1 | - | 5 - 6 | 676 | 4913757 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |