Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007758.2 | NP_031784.1 | 1 - 2 | 160 | 66 | |
GenBank mRNA | AK003072.2 | - | 1 - 2 | 160 | 66 | |
BC109344.1 | - | 1 - 2 | 93 | 40054528 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |