Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282054.1 | NP_001268983.1 | 8 - 9 | 1289 | 61 | |
NM_031870.3 | NP_114076.1 | 7 - 8 | 1380 | 61 | ||
GenBank mRNA | AF178957.1 | - | 7 - 8 | 1084 | 61 | |
AF298655.1 | - | 7 - 8 | 1380 | 61 | ||
AK029837.1 | - | 8 - 9 | 1290 | 61 | ||
AK161474.1 | - | 7 - 8 | 1130 | 61 | ||
AY351585.1 | - | 7 - 8 | 1334 | 61 | ||
BC138163.1 | - | 7 - 8 | 1271 | 8861182 | 61 | |
BC145838.1 | - | 7 - 8 | 1271 | 40131141 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |