Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tet2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040400.2 | NP_001035490.2 | 197 | 130 | ||
GenBank mRNA | AK155776.1 | - | 1 - 2 | 110 | 130 | |
BF463367.1 | - | 1 - 2 | 197 | 130 | ||
DQ079067.1 | - | 1 - 2 | 175 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |