Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpe65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029987.2 | NP_084263.2 | 1 - 2 | 59 | 109 | |
GenBank mRNA | AK020906.1 | - | 1 - 2 | 58 | 109 | |
CJ133618.1 | - | 1 - 2 | 60 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Inventoried Probe spans exons |