Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK173191.1 | - | 11 - 12 | 5497 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |