Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027808.1 | NP_082084.1 | 7 - 8 | 1178 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |