Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026582.4 | NP_080858.3 | 10 - 11 | 1604 | 71 | |
NR_037590.1 | 11 - 12 | 1685 | 71 | |||
GenBank mRNA | AK019886.1 | - | 11 - 12 | 1643 | 71 | |
AK029053.1 | - | 10 - 11 | 1563 | 71 | ||
AK029104.1 | - | 10 - 11 | 1561 | 71 | ||
AK044810.1 | - | 10 - 11 | 1563 | 71 | ||
AK049842.1 | - | 10 - 11 | 1561 | 71 | ||
AK077407.1 | - | 10 - 11 | 1562 | 71 | ||
AK128912.1 | - | 10 - 11 | 1560 | 71 | ||
AK159207.1 | - | 10 - 11 | 1561 | 71 | ||
BC018381.1 | - | 9 - 10 | 1188 | 4237619 | 71 | |
BC075615.1 | - | 10 - 11 | 1604 | 5703610 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |