Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Furin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081454.1 | NP_001074923.1 | 12 - 13 | 1647 | 63 | |
NM_011046.2 | NP_035176.1 | 12 - 13 | 1767 | 63 | ||
GenBank mRNA | BC048234.1 | - | 11 - 12 | 1474 | 6492009 | 63 |
L26489.1 | - | 12 - 13 | 1767 | 63 | ||
X54056.1 | - | 11 - 12 | 1470 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |