Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300011794.mm.1 |
NM_053102.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011794.mm.1 |
NM_053102.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011794.mm.1 |
TC0300001469.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300011788.mm.1 |
NM_053102.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011788.mm.1 |
NM_053102.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011788.mm.1 |
TC0300001469.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300011793.mm.1 |
NM_053102.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011793.mm.1 |
NM_053102.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011793.mm.1 |
TC0300001469.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300011790.mm.1 |
NM_053102.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011790.mm.1 |
NM_053102.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011790.mm.1 |
TC0300001469.mm.1 |
MTA TC |