Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133871.2 | NP_598632.2 | 4 - 5 | 858 | 104 | |
GenBank mRNA | AJ299405.1 | - | 3 - 4 | 748 | 104 | |
AK079749.1 | - | 4 - 5 | 810 | 104 | ||
AK136553.1 | - | 4 - 5 | 810 | 104 | ||
AK151346.1 | - | 4 - 5 | 809 | 104 | ||
AK171676.1 | - | 4 - 5 | 791 | 104 | ||
BC111882.1 | - | 3 - 4 | 748 | 40060245 | 104 | |
BC112402.1 | - | 3 - 4 | 748 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |