Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ska3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198605.3 | NP_941007.1 | 4 - 5 | 798 | 156 | |
GenBank mRNA | AK089223.1 | - | 4 - 5 | 799 | 156 | |
AK158142.1 | - | 4 - 5 | 820 | 156 | ||
AK166171.1 | - | 4 - 5 | 789 | 156 | ||
BC117500.1 | - | 4 - 5 | 790 | 40105921 | 156 | |
BC117501.1 | - | 4 - 5 | 790 | 40105920 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |