Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ska3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198605.3 | NP_941007.1 | 5 - 6 | 899 | 70 | |
GenBank mRNA | AK089223.1 | - | 5 - 6 | 900 | 70 | |
AK158142.1 | - | 5 - 6 | 921 | 70 | ||
AK166171.1 | - | 5 - 6 | 890 | 70 | ||
BC117500.1 | - | 5 - 6 | 891 | 40105921 | 70 | |
BC117501.1 | - | 5 - 6 | 891 | 40105920 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |