Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Loxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033325.2 | NP_201582.2 | 9 - 10 | 1777 | 124 | |
GenBank mRNA | AK034957.1 | - | 9 - 10 | 1815 | 124 | |
AK045019.1 | - | 9 - 10 | 1777 | 124 | ||
AK145323.1 | - | 9 - 10 | 1815 | 124 | ||
AK159386.1 | - | 9 - 10 | 1811 | 124 | ||
BC086801.1 | - | 2059 | 30605240 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |