Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mttp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC012686.1 | - | 6 - 7 | 794 | 4163901 | 76 |
EU553486.1 | - | 6 - 7 | 802 | 76 | ||
L47970.1 | - | 6 - 7 | 781 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |