Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qpctl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026111.3 | NP_080387.2 | 1 - 2 | 219 | 120 | |
GenBank mRNA | AK007555.1 | - | 1 - 2 | 267 | 120 | |
AK007701.1 | - | 1 - 2 | 247 | 120 | ||
AK035587.1 | - | 1 - 2 | 219 | 120 | ||
AK045687.1 | - | 1 - 2 | 223 | 120 | ||
BC058181.1 | - | 1 - 2 | 242 | 5325516 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |