Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012058.3 | NP_036188.1 | 2 - 3 | 311 | 87 | |
GenBank mRNA | AK011720.1 | - | 2 - 3 | 239 | 87 | |
AK020583.1 | - | 2 - 3 | 260 | 87 | ||
AK020620.1 | - | 2 - 3 | 252 | 87 | ||
AK051208.1 | - | 2 - 3 | 257 | 87 | ||
AK146882.1 | - | 2 - 3 | 260 | 87 | ||
AK169314.1 | - | 2 - 3 | 259 | 87 | ||
BC039648.1 | - | 2 - 3 | 298 | 5353755 | 87 | |
BC090254.1 | - | 2 - 3 | 239 | 5100716 | 87 | |
X78304.1 | - | 2 - 3 | 237 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |