Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc42bpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033285.1 | NP_001028457.1 | 8 - 9 | 1167 | 93 | |
GenBank mRNA | AK138649.1 | - | 9 - 10 | 1656 | 93 | |
BC158017.1 | - | 8 - 9 | 1167 | 9007323 | 93 | |
BC158095.1 | - | 8 - 9 | 1167 | 9007382 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |