Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010713.2 | NP_034843.2 | 2 - 3 | 373 | 126 | |
GenBank mRNA | AK160121.1 | - | 2 - 3 | 374 | 126 | |
AK165733.1 | - | 2 - 3 | 367 | 126 | ||
BC125281.1 | - | 2 - 3 | 282 | 40129796 | 126 | |
BC125283.1 | - | 2 - 3 | 282 | 40129798 | 126 | |
BC144768.1 | - | 2 - 3 | 282 | 9053316 | 126 | |
D49658.1 | - | 2 - 3 | 342 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |