Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syde2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166064.1 | NP_001159536.1 | 4 - 5 | 3160 | 104 | |
GenBank mRNA | AK049496.1 | - | 4 - 5 | 2378 | 104 | |
CN702275.1 | - | 2 - 3 | 216 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |