Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085472.2 | NP_001078941.2 | 9 - 10 | 947 | 69 | |
NM_001085473.2 | NP_001078942.2 | 15 - 16 | 3272 | 69 | ||
NM_001242605.1 | NP_001229534.1 | 8 - 9 | 1011 | 69 | ||
NM_001242606.1 | NP_001229535.1 | 8 - 9 | 972 | 69 | ||
NM_019567.3 | NP_062513.3 | 9 - 10 | 1076 | 69 | ||
NM_023190.3 | NP_075679.2 | 15 - 16 | 3308 | 69 | ||
GenBank mRNA | AF124725.1 | - | 9 - 10 | 1007 | 69 | |
AF124729.1 | - | 8 - 9 | 942 | 69 | ||
AF168782.1 | - | 15 - 16 | 3309 | 69 | ||
AK012099.1 | - | 9 - 10 | 940 | 69 | ||
AK030449.1 | - | 15 - 16 | 3002 | 69 | ||
AK122342.1 | - | 15 - 16 | 2943 | 69 | ||
BC019127.1 | - | 8 - 9 | 996 | 69 | ||
BC052755.1 | - | 8 - 9 | 932 | 69 | ||
BC075691.1 | - | 15 - 16 | 2957 | 69 | ||
BC094217.1 | - | 8 - 9 | 954 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |