Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028665.4 | NP_082941.2 | 6 - 7 | 1747 | 64 | |
GenBank mRNA | AK029511.1 | - | 6 - 7 | 1749 | 64 | |
AK029888.1 | - | 6 - 7 | 1708 | 64 | ||
AK033599.1 | - | 6 - 7 | 1736 | 64 | ||
AK048925.1 | - | 6 - 7 | 1641 | 64 | ||
AK133339.1 | - | 1708 | 64 | |||
BC030885.1 | - | 6 - 7 | 1376 | 64 | ||
BC100433.1 | - | 6 - 7 | 1783 | 64 | ||
BC145456.1 | - | 6 - 7 | 865 | 9054017 | 64 | |
BC145457.1 | - | 5 - 6 | 729 | 64 | ||
BC145458.1 | - | 5 - 6 | 729 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |