Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028243.3 | NP_082519.1 | 2 - 3 | 391 | 107 | |
GenBank mRNA | AK011112.1 | - | 2 - 3 | 373 | 107 | |
AK051677.1 | - | 2 - 3 | 348 | 107 | ||
AK054462.1 | - | 2 - 3 | 329 | 107 | ||
AK166282.1 | - | 2 - 3 | 348 | 107 | ||
BC026424.1 | - | 2 - 3 | 187 | 107 | ||
BC055022.1 | - | 2 - 3 | 324 | 4017462 | 107 | |
BC089575.1 | - | 2 - 3 | 346 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |