Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023733.3 | NP_076222.1 | 9 - 10 | 1033 | 63 | |
GenBank mRNA | AK004567.2 | - | 9 - 10 | 1034 | 63 | |
BC006593.1 | - | 9 - 10 | 1021 | 3156501 | 63 | |
BC012308.1 | - | 9 - 10 | 1048 | 3500167 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |