Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033314.3 | NP_001028486.1 | 7 - 8 | 1123 | 61 | |
GenBank mRNA | AK146297.1 | - | 7 - 8 | 1124 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |